Order Entry
Austria
Orders LinkContactUsLinkComponent
Buffer solutions

Buffer solutions

Type L4790, 4,01/6,87, ±0,010, 25 °C, 60 Ampoul
Catalog # 662-1608
Supplier:   SI Analytics
Buffer solutions
Catalog # 662-1608
Supplier:  SI Analytics
CAS Number:  
Buffer solutions
Buffer solutions

Type L4790, 4,01/6,87, ±0,010, 25 °C, 60 Ampoul

CAS Number:  
Price
261,00 €
Log in for availability
1 * 60 Ampull
Product Details & Documents

Buffer solutions intended for extremely precise calibration.

  • Highest measurement reliability
  • No preservative agents
  • Hermetically sealed in the glass ampoule
  • Sterilised with hot steam

Certifications: According to DIN 19 266.

Specifications

Specifications

  • Pk:
    60 Ampoul
  • Reference temperature:
    25 °C
  • Description:
    Type L4790
  • pH accuracy:
    ±0,010
  • pH value:
    4,01/6,87

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.