Order Entry
Austria
Orders LinkContactUsLinkComponent
Buffer solutions

Buffer solutions

Type L7004, 7,00, ±0,010, 25 °C, 250 mL
Catalog # 662-6175
Supplier:   SI Analytics
Buffer solutions
Catalog # 662-6175
Supplier:  SI Analytics
CAS Number:  
Buffer solutions
Buffer solutions

Type L7004, 7,00, ±0,010, 25 °C, 250 mL

CAS Number:  
Price
36,40 €
Log in for availability
1 * 250 mL
Product Details & Documents

Buffer solutions intended for extremely precise calibration.

  • Highest measurement reliability
  • No preservative agents
  • Hermetically sealed in the glass ampoule
  • Sterilised with hot steam

Certifications: According to DIN 19 266.

Specifications

Specifications

  • Pk:
    250 mL
  • Reference temperature:
    25 °C
  • Description:
    Type L7004
  • pH accuracy:
    ±0,010
  • pH value:
    7,00

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.