Electrode Cables
BNC connector for pH meters with BNC for example Hach, Orion, Mettler, 1 m
Catalog # 662-0062
Supplier: SI Analytics
Jump to:
Product Details & Documents
Electrode cables with a range of connectors to fit most pH meters.
Specifications
- Description:BNC connector for pH meters with BNC for example Hach, Orion, Mettler
- Length:1 m
- Seq:16