Orders LinkContactUsLinkComponent
VWR®, Calibration Weights, Class M2
VWR®, Calibration Weights, Class M2
10 kg, 1600 ± mg
Catalog # 611-8382
Supplier:   VWR Collection
VWR®, Calibration Weights, Class M2
Catalog # 611-8382
Supplier:  VWR Collection
VWR®, Calibration Weights, Class M2
VWR®, Calibration Weights, Class M2
10 kg, 1600 ± mg
Price
861,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Test weights in either stainless steel or aluminium.
Delivery information: Supplied in a plastic box.
Specifications
  • Weight:
    10 kg
  • Seq:
    16
  • Tolerance:
    1600 ± mg