Order Entry
Austria
Orders LinkContactUsLinkComponent
Separating funnels, Squibb, Nalgene®

Separating funnels, Squibb, Nalgene®

1000 ml, 53 mm
Catalog # 527-0008
   Low Manufacturing Impact
Separating funnels, Squibb, Nalgene®
Catalog # 527-0008
   Low Manufacturing Impact
Separating funnels, Squibb, Nalgene®
Separating funnels, Squibb, Nalgene®

1000 ml, 53 mm

Price
141,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

PPCO translucent, TFE stopcock, PP screw closure.

  • Leakproof closure and stopcock
  • Very good chemical resistance
  • Autoclavable except for stopcock assembly
  • TFE stopcock requires no lubrication
Specifications
  • Capacity:
    1000 ml
  • Ø cap:
    53 mm
  • Cat. No.:
    527-0008
  • Environmental certification:
    NAR Certificate of Retirement
  • Environmentally Preferable:
    Low Manufacturing Impact
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.