Order Entry
Austria
Orders LinkContactUsLinkComponent
Separating funnels, Squibb, Nalgene®

Separating funnels, Squibb, Nalgene®

500 ml, 43 mm
Catalog # 527-0007
   Low Manufacturing Impact
Separating funnels, Squibb, Nalgene®
Catalog # 527-0007
   Low Manufacturing Impact
Separating funnels, Squibb, Nalgene®
Separating funnels, Squibb, Nalgene®

500 ml, 43 mm

Price
113,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

PPCO translucent, TFE stopcock, PP screw closure.

  • Leakproof closure and stopcock
  • Very good chemical resistance
  • Autoclavable except for stopcock assembly
  • TFE stopcock requires no lubrication
Specifications
  • Capacity:
    500 ml
  • Ø cap:
    43 mm
  • Cat. No.:
    527-0007
  • Environmental certification:
    NAR Certificate of Retirement
  • Environmentally Preferable:
    Low Manufacturing Impact