Order Entry
Austria
Orders LinkContactUsLinkComponent
Büchner funnels, Nalgene®

Büchner funnels, Nalgene®

186 ml, 70 mm
Catalog # 221-0101
   Low Manufacturing Impact
Büchner funnels, Nalgene®
Catalog # 221-0101
   Low Manufacturing Impact
Büchner funnels, Nalgene®
Büchner funnels, Nalgene®

186 ml, 70 mm

Price
19,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

PP

  • Robust and lightweight
  • Top and bottom can be separated for easy cleaning
  • Autoclavable

For filter paper or membrane filtration discs.

Specifications
  • Capacity:
    186 ml
  • For filter Ø:
    70 mm
  • Cat. No.:
    221-0101
  • Environmental certification:
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • Environmentally Preferable:
    Low Manufacturing Impact
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.