Orders LinkContactUsLinkComponent
Volumetric flasks, class B, Nalgene®
Volumetric flasks, class B, Nalgene®
100 ml, 0,16 ± ml
Catalog # 612-3022
   Low Manufacturing Impact
Volumetric flasks, class B, Nalgene®
Catalog # 612-3022
   Low Manufacturing Impact
Volumetric flasks, class B, Nalgene®
Volumetric flasks, class B, Nalgene®
100 ml, 0,16 ± ml
Price
53,40 €
Log in for availability
1 * 1 ST
Product Details & Documents
PMP (TPX®), transparent, with PP screw cap.
  • Very robust and allowing precise measurement
  • Calibrated individually with permanent colour graduation line
  • Autoclavable when the stopper has been removed from the threaded neck and a small quantity of water has been added to the flask. Cleaning with mild alkaline detergents recommended up to +60 °C (accuracy may be affected over +60 °C)
ASTM E288 and ISO Standard 384 Class B
 : ASTM E288 and ISO Standard 384 Class B
Specifications
  • Capacity:
    100 ml
  • Tolerance:
    0,16 ± ml
  • Cat. No.:
    612-3022
  • Environmental certification:
    NAR Certificate of Retirement
  • Environmentally Preferable:
    Low Manufacturing Impact