Order Entry
Austria
Orders LinkContactUsLinkComponent
Tanks, rectangular, with cover, Nalgene®

Tanks, rectangular, with cover, Nalgene®

20 L, 356×250×250 mm
Catalog # 216-1922
   Low Manufacturing Impact
Tanks, rectangular, with cover, Nalgene®
Catalog # 216-1922
   Low Manufacturing Impact
Tanks, rectangular, with cover, Nalgene®
Tanks, rectangular, with cover, Nalgene®

20 L, 356×250×250 mm

Price
593,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

These PP rectangular tanks with cover can significantly reduce evaporation and contamination.

  • Good stress-crack resistance
  • Suitable for use with many organic chemicals
  • Tanks can be modified with a factory-installed spigot
Specifications
  • Capacity:
    20 L
  • Seq:
    02
  • W×D×H:
    356×250×250 mm
  • Cat. No.:
    216-1922
  • Environmental certification:
    NAR Certificate of Retirement
  • Environmentally Preferable:
    Low Manufacturing Impact