pH Papers, without Colour Scale
pH test paper Congo paper (red → blue) MN 260 HE, 200 test strips, length 70 mm and width 20 mm, 5,0 - 3,0, Plastic box, 48×25×84 mm, 47,4 g
Anbieter: MACHEREY-NAGEL
Jump to:
Product Details & Documents
Easy to use pH papers to control pH adjustments.
Long term stable and protected against external influences due to plastic packaging
- No accessories needed - always ready to use
- Quick and easy determination of the pH
These products are simple, completely impregnated indicator papers, which do not allow exact pH determinations, but pH adjustments during special chemical procedures. For this reason, no colour scale is provided with these papers.
Certifications: Not CE certified.
Caution: Contains hazardous materials: MANA90703 and MANA90705.
Documents
Specifications
- pH range:5,0 - 3,0
- Description:pH test paper Congo paper (red → blue) MN 260 HE, 200 test strips, length 70 mm and width 20 mm
- Seq:3
- Pack type:Plastic box, 48×25×84 mm, 47,4 g
- Cat. No.:MANA90705
- Shelf life:2 year(s)
Empfehlungen
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen.



