Order Entry
Austria
Orders LinkContactUsLinkComponent
Extraction thimbles

Extraction thimbles

75×160 mm, 2,0 mm
Catalog # MANA645041
Supplier:   MACHEREY-NAGEL
Extraction thimbles
Catalog # MANA645041
Supplier:  MACHEREY-NAGEL
Extraction thimbles
Extraction thimbles

75×160 mm, 2,0 mm

Price
294,00 €
Log in for availability
1 * 25 ST
Product Details & Documents

Extraction thimbles, made of pure cellulose, standard quality. With or without lid available.

  • Fat determination in food and feed
  • Determination of binder content in asphalt
  • Analysis of soil contaminants
  • Determination of additives in rubber and elastomers
Specifications
  • Int.Ø×H:
    75×160 mm
  • Seq:
    43
  • Thickness:
    2,0 mm