Order Entry
Austria
Orders LinkContactUsLinkComponent
Halogen bulbs

Halogen bulbs

Halogen bulb, 12 V, 20 W
Catalog # 632-8963
Supplier:   MOTIC
Halogen bulbs
Catalog # 632-8963
Supplier:  MOTIC
Halogen bulbs
Halogen bulbs

Halogen bulb, 12 V, 20 W

Price
11,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    Halogen bulb
  • Seq:
    01
  • Power:
    20 W
  • Voltage:
    12 V
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.