ContactUsLinkComponent
REFRIGERATOR FORCED AIR COOLING 352 L HEIGHT 1590 X WIDTH 602 X DEPTH 600 MM 5 SHELVES AND 1 GRID ENERGY CONSUMPTION 1.60 KWH/24 HOU RS 1 * 1 items
Catalog # 472-2204
Supplier:  Liebherr
REFRIGERATOR FORCED AIR COOLING 352 L HEIGHT 1590 X WIDTH 602 X DEPTH 600 MM 5 SHELVES AND 1 GRID ENERGY CONSUMPTION 1.60 KWH/24 HOU RS 1 * 1 items
Catalog # 472-2204
Supplier:  Liebherr
Frequently Bought Together