Orders LinkContactUsLinkComponent
Adapters, bends
Adapters, bends
19/26 NS
Catalog # LENZ05020002
Supplier:   Lenz Laborglas
Adapters, bends
Catalog # LENZ05020002
Supplier:  Lenz Laborglas
Adapters, bends
Adapters, bends
19/26 NS
Price
25,30 €
Log in for availability
1 * 1 ST
Product Details & Documents
DURAN®, borosilicate glass, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Inclined at an angle of 75°. With two standard ground cones.
Documents
Specifications
  • Cone size:
    19/26 NS