Multiple adapters
29/32 NS, 29/32 NS
Catalog # 201-2221
Supplier: Lenz Laborglas
Jump to:
Product Details & Documents
DURAN® Borosilicate glass 3.3
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
With 3 NS sockets and 1 NS cone, with two parallel necks and one angled neck (45°).
Documents
BSE TSE Glass Columns
BSE/TSE Certificates
Specifications
- Cone size:29/32 NS
- Socket size:29/32 NS