Orders LinkContactUsLinkComponent
Multiple adapters
Multiple adapters
29/32 NS, 29/32 NS
Catalog # 201-2216
Supplier:   Lenz Laborglas
Multiple adapters
Catalog # 201-2216
Supplier:  Lenz Laborglas
Multiple adapters
Multiple adapters
29/32 NS, 29/32 NS
Price
60,70 €
Log in for availability
1 * 1 ST
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Two sockets and one cone, standard ground joints and two parallel necks.
Documents
Specifications
  • Cone size:
    29/32 NS
  • Socket size:
    29/32 NS