Conical separating funnels
250 ml, 29/32 NS, -
Artikelnr. 527-1133
Anbieter: Lenz Laborglas
Springe zu:
Product Details & Documents
DURAN®, borosilicate glass, with NS-PE stopper.
With socket and plain stem. Available with interchangeable solid glass key or alternatively PTFE key for grease-free performance. Both with or without graduations.
Dokumente
BSE TSE Glass Columns
BSE/TSE Certificates
LENZ04000073_TDS_EN
Datasheet
Spezifikationen
- Socket size:29/32 NS
- Capacity:250 ml
- Division:-