Stopcocks, three way
1,5 mm, 14,5 NS
227-3231
DURAN®, borosilicate glass, with solid glass or PTFE key.
- Replaceable NS keys with T-bore
- Screw thread retaining device
- Leakage rate compliant with DIN 12540-1
DIN 12554
- Hole Ø:1,5 mm
- Spigot:14,5 NS