Orders LinkContactUsLinkComponent
Stopcocks, two-way
Stopcocks, two-way
2,5 mm, 14,5 NS
Catalog # LENZ02431402
Supplier:   Lenz Laborglas
Stopcocks, two-way
Catalog # LENZ02431402
Supplier:  Lenz Laborglas
Stopcocks, two-way
Stopcocks, two-way
2,5 mm, 14,5 NS
Price
77,40 €
Log in for availability
1 * 1 ST
Product Details & Documents
Stopcocks are made of DURAN® borosilicate glass with solid NS glass plug, screwthread retaining device, and two oblique bores.
  • Excellent chemical resistance
  • Excellent temperature resistance
With two oblique bores.
Leakage rate accoring to DIN 12540-1.
Documents
Specifications
  • Hole Ø:
    2,5 mm
  • Spigot:
    14,5 NS