Order Entry
Austria
Orders LinkContactUsLinkComponent
Bath fluids

Bath fluids

Therm 180*, 0...180 °C, >288 °C, >252 °C, Colourless, 5 l
Catalog # 461-2848
Supplier:   LAUDA
Bath fluids
Catalog # 461-2848
Supplier:  LAUDA
CAS Number:  
Bath fluids
Bath fluids

Therm 180*, 0...180 °C, >288 °C, >252 °C, Colourless, 5 l

CAS Number:  
Price
469,00 €
Log in for availability
1 * 5 L
Product Details & Documents

Allow highly accurate thermostating, even at extreme temperatures.

  • Simple and safe handling, suitable for long term operation
  • Optimise thermostat life

* Denotes Silicone oil
** Only suitable for closed circuits

Specifications

Specifications

  • Pk:
    5 l
  • Colour:
    Colourless
  • Fire point:
    >288 °C
  • Flash point:
    >252 °C
  • Temperature range:
    0...180 °C
  • Type:
    Therm 180*

Specifications