Dewar flasks, type GSS
Loose lid insulating plug, GSS 1000 W + GSS 2000
478-5144
: KGW
Dewar flasks in stainless steel for storage and transport of liquid nitrogen, dry ice and other coolants. They are vacuum insulated and have a temperature range from −200 to +200 °C.
- With side grip and transport handle (except model GSS 500)
- Description:Loose lid insulating plug
- Seq:09
- For:GSS 1000 W + GSS 2000
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



