Orders LinkContactUsLinkComponent
Filter discs
Filter discs
90 mm
Catalog # 519-5005
Supplier:   KARTELL
Filter discs
Catalog # 519-5005
Supplier:  KARTELL
Filter discs
Filter discs
90 mm
Price
15,10 €
Log in for availability
1 * 10 ST
Product Details & Documents
HDPE filter discs for use between filter paper and perforated Büchner plate to prevent clogging of filter holes.
Specifications
  • Ø:
    90 mm
  • Seq:
    05