Order Entry
Austria
Orders LinkContactUsLinkComponent
Press-down stoppers

Press-down stoppers

Natural, 38 mm, 40 mm
Catalog # 212-0217
Supplier:   KARTELL
Press-down stoppers
Catalog # 212-0217
Supplier:  KARTELL
Press-down stoppers
Press-down stoppers

Natural, 38 mm, 40 mm

Price
25,30 €
Log in for availability
1 * 100 ST
Product Details & Documents

For sealing glass or plastic reagent and sample tubes.

  • PE, natural colour
  • Good chemical resistance
  • Economical
  • Convenient pack sizes

*conical

Specifications
  • Ø bottom:
    38 mm
  • Ø top:
    40 mm
  • Colour:
    Natural