Press-down stoppers
Natural, 33 mm, 35 mm
Catalog # KART657
Supplier: KARTELL
Jump to:
Product Details & Documents
For sealing glass or plastic reagent and sample tubes.
- PE, natural colour
- Good chemical resistance
- Economical
- Convenient pack sizes
*conical
Specifications
- Ø bottom:33 mm
- Ø top:35 mm
- Colour:Natural



