Press-down stoppers
Natural, 10,5 mm, 12 mm
Catalog # 212-0212
Supplier: KARTELL
Jump to:
Product Details & Documents
For sealing glass or plastic reagent and sample tubes.
- PE, natural colour
- Good chemical resistance
- Economical
- Convenient pack sizes
*conical
Specifications
- Ø bottom:10,5 mm
- Ø top:12 mm
- Colour:Natural
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



