Order Entry
Austria
Orders LinkContactUsLinkComponent
Büchner funnels

Büchner funnels

180 ml, 70 mm
Catalog # 519-2363
Supplier:   KARTELL
Büchner funnels
Catalog # 519-2363
Supplier:  KARTELL
Büchner funnels
Büchner funnels

180 ml, 70 mm

Price
27,20 €
Log in for availability
1 * 1 ST
Product Details & Documents

PP, white.

  • Robust and lightweight
  • Top and bottom can be separated for easy cleaning

For filter paper or membrane filtration discs.

Specifications
  • Capacity:
    180 ml
  • For filter Ø:
    70 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.