Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Tissue embedding cassettes, UNI type

Blue
  631-9611
 :   KARTELL
Tissue embedding cassettes, UNI type
  631-9611
 :  KARTELL
Tissue embedding cassettes, UNI type
Tissue embedding cassettes, UNI type

Blue

 
95,90 €
   
1 * 500 ST
 

Tissue embedding cassettes for paraffin inclusions, Universal type, provided with clip on closure lid that can easily be removed. They are available in five colours. Dimensions: 28×40×6,8 mm.

 
  • Colour:
    Blue
Accessories

 

  •  
    Description
     
     
  • KART2922-04
    Tissue embedding sponge, blue
       
    1 * 500 ST - 29,40 €