Order Entry
Austria
Orders LinkContactUsLinkComponent
Tissue embedding cassettes, UNI type

Tissue embedding cassettes, UNI type

White
Catalog # 631-9610
Supplier:   KARTELL
Tissue embedding cassettes, UNI type
Catalog # 631-9610
Supplier:  KARTELL
Tissue embedding cassettes, UNI type
Tissue embedding cassettes, UNI type

White

Price
95,90 €
Log in for availability
1 * 500 ST
Product Details & Documents

Tissue embedding cassettes for paraffin inclusions, Universal type, provided with clip on closure lid that can easily be removed. They are available in five colours. Dimensions: 28×40×6,8 mm.

Specifications
  • Colour:
    White
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Tissue embedding sponge, blue
    Tissue embedding sponge, blue
    KART2922-04
    Log in for availability
    1 * 500 ST - 29,40 €