Order Entry
Austria
Orders LinkContactUsLinkComponent
Slide boxes

Slide boxes

Grey, PS, 100, 280×230×35 mm
Catalog # 631-9142
Supplier:   KARTELL
Slide boxes
Catalog # 631-9142
Supplier:  KARTELL
Slide boxes
Slide boxes

Grey, PS, 100, 280×230×35 mm

Price
67,30 €
Log in for availability
1 * 1 ST
Product Details & Documents

High impact resistant slide boxes with removable lids.

  • Well spaced slots for easy handling of slides
  • Numbering for cross reference with corresponding index card
  • Locating lugs so boxes can be stacked

W×D×H: 83×98×38 mm (for 25); 97×230×35 mm (for 50); 280×230×35 mm (for 100)

Specifications
  • No. of slides:
    100
  • Colour:
    Grey
  • Material:
    PS
  • W×D×H:
    280×230×35 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.