Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Volumetric flasks, class B

25 ml, 12/21, 121 mm
  612-3000
 :   KARTELL
Volumetric flasks, class B
  612-3000
 :  KARTELL
Volumetric flasks, class B
Volumetric flasks, class B

25 ml, 12/21, 121 mm

 
14,40 €
   
1 * 1 ST
 

PP, translucent, with PE stopper.

  • Narrow neck with no meniscus for easy reading
  • Calibrated gravimetrically at +20 °C
  • Autoclavable, chemically non absorbent, non wetting surface
 
  • NS:
    12/21
  • Capacity:
    25 ml
  • Height:
    121 mm