Orders LinkContactUsLinkComponent
Cap inserts, PP, Corning®
Cap inserts, PP, Corning®
Red
Artikelnr. 479-3002
   Low Manufacturing Impact
Anbieter:   Corning
Cap inserts, PP, Corning®
Artikelnr. 479-3002
Anbieter:  Corning
   Low Manufacturing Impact
Cap inserts, PP, Corning®
Cap inserts, PP, Corning®
Red
Preis
46,10 €
Log in for availability
1 * 500 ST
Product Details & Documents
The white polypropylene cap insert provide colour-coding for easy sample identification.
  • Inserts are packaed in resealable bags
  • Non-sterile
Dokumente
Specifications
  • Colour:
    Red
  • Seq:
    23
  • Cat. No.:
    479-3002
  • Environmental certification:
    I-RECs, ISO 14001 - Reynosa
  • Environmentally Preferable:
    Low Manufacturing Impact