Orders LinkContactUsLinkComponent
Test sets
Test sets
Test set I for inorganic gases
Catalog # 151-0176
Supplier:   Dräger
Test sets
Catalog # 151-0176
Supplier:  Dräger
Test sets
Test sets
Test set I for inorganic gases
Price
84,50 €
Log in for availability
1 * 1 ST
Product Details & Documents
These test sets allow for the semi quantitative measurements of five different gases at the same time.
Specifications
  • Description:
    Test set I for inorganic gases
  • Seq:
    01
  • Cat. No.:
    151-0176
  • Model:
    Test set I for inorganic gases
  • Measurement time:
    40 s