Test sets
Adapter for simultaneous test via connection to a bellows pump
Catalog # 151-0116
Supplier: Dräger
Jump to:
Product Details & Documents
These test sets allow for the semi quantitative measurements of five different gases at the same time.
Specifications
- Description:Adapter for simultaneous test via connection to a bellows pump
- Seq:07