Sodium fluoride 99+% for analysis
Plastic bottle, 25 g
Supplier: Thermo Fisher Scientific
Sodium fluoride 99+% for analysis
Catalog # ACRO201290250
Supplier: Thermo Fisher Scientific
CAS Number: 7681-49-4
Jump to:
Product Details & Documents
- CAS Number:7681-49-4
- Formula:NaF
- MDL Number:MFCD00003524
- Molecular Weight:41.99 g/mol
- Boiling Pt:1700 °C (1013 hPa)
- Melting Pt:993 °C
- Density:2,8 g/cm³ (20 °C)
- Storage Temperature:Ambient
- UN:1690
- ADR:6.1,III
Documents
ACRO20129
SDS
Specifications
Specification Test Results
- Appearance:White Powder
- Loss on drying:≤0.2 % (150°C)
- Titration:≥99.0 %
- Chloride (Cl):≤50 ppm
- Sulfate (SO4):≤1000 ppm
- Sodium fluosilicate:≤0.1 %
- Iron (Fe):≤50 ppm
- Heavy metals (as Pb):≤10 ppm
- Free acid (HF):≤0.1 %
- Free alkali (NaOH):≤0.1 %
Specifications
- Pack type:Plastic bottle
- Pk:25 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



