Orders LinkContactUsLinkComponent
Stopcocks, straight through, with tip
Stopcocks, straight through, with tip
PP
Catalog # 661-1183
Supplier:   COWIE
Stopcocks, straight through, with tip
Catalog # 661-1183
Supplier:  COWIE
Stopcocks, straight through, with tip
Stopcocks, straight through, with tip
PP
Price
16,10 €
Log in for availability
1 * 10 ST
Product Details & Documents
Made of PTFE.
  • Available with PP or glass tip
  • Very good chemical resistance
  • Very good temperature resistance
Specifications
  • Description:
    PP