ContactUsLinkComponent
GLOVES CONFORM + : AMBIDEXTROUS SINGLE-USE LATEX GLOVES. PRODUCED UNDER ISO 9002. POWDER FREE. TEXTURED FINISH. NATURAL COLOUR. SIZ E 8.5 1 * 100 items
  112-0977
 :  Ansell
GLOVES CONFORM + : AMBIDEXTROUS SINGLE-USE LATEX GLOVES. PRODUCED UNDER ISO 9002. POWDER FREE. TEXTURED FINISH. NATURAL COLOUR. SIZ E 8.5 1 * 100 items
  112-0977
 :  Ansell
Frequently Bought Together