ContactUsLinkComponent
GLOVES SOL-VEX : UNSUPPORTED NITRILE. PRODUCED UNDER ISO 9002. GREEN. UNFLOCKED. THICKNESS 0.56 MM.LENGTH 380 MM. REVERSED LOZENGE. SIZE 11 1 * 12 PAIR
Catalog # 112-0285
Supplier:  Ansell
GLOVES SOL-VEX : UNSUPPORTED NITRILE. PRODUCED UNDER ISO 9002. GREEN. UNFLOCKED. THICKNESS 0.56 MM.LENGTH 380 MM. REVERSED LOZENGE. SIZE 11 1 * 12 PAIR
Catalog # 112-0285
Supplier:  Ansell
Frequently Bought Together