Orders LinkContactUsLinkComponent
Thermometer test caps
TX25L, test cap, Lumberg type, +63 °C
Catalog # 620-0379
Supplier:   COMARK
Thermometer test caps
Catalog # 620-0379
Supplier:  COMARK
Thermometer test caps
Thermometer test caps
TX25L, test cap, Lumberg type, +63 °C
Price
144,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Certified test capsules can be used for quick checking of the thermometer's function against specifications. They are connected to the thermometer instead of a sensor and consist of a resistor that corresponds exactly to a given temperature.
Specifications
  • Range:
    +63 °C
  • Description:
    TX25L, test cap, Lumberg type