Order Entry
Austria
Orders LinkContactUsLinkComponent
Thermometer probes

Thermometer probes

Type K industrial probe, with sub-miniature connector, IK21M, −100...+850 °C, 1,5×100 mm
Catalog # 620-3171
Supplier:   COMARK
Thermometer probes
Catalog # 620-3171
Supplier:  COMARK
Thermometer probes

Type K industrial probe, with sub-miniature connector, IK21M, −100...+850 °C, 1,5×100 mm

Price
272,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Range:
    −100...+850 °C
  • Description:
    Type K industrial probe, with sub-miniature connector, IK21M
  • Application:
    Immersion
  • Ø×L:
    1,5×100 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.