Orders LinkContactUsLinkComponent
Thermometer probes
Thermometer probes
Flexible wire type K air probe, with sub-miniature connector, AK29M, −100…+250 °C, - mm
Catalog # COMAAK29M
Supplier:   COMARK
Thermometer probes
Catalog # COMAAK29M
Supplier:  COMARK
Thermometer probes
Flexible wire type K air probe, with sub-miniature connector, AK29M, −100…+250 °C, - mm
Price
80,50 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Range:
    −100…+250 °C
  • Description:
    Flexible wire type K air probe, with sub-miniature connector, AK29M
  • Ø×L:
    - mm