Order Entry
Austria
Orders LinkContactUsLinkComponent
Thermometer probes

Thermometer probes

Mineral insulated semi flexible type K probe, with sub-miniature connector, AK21M, −100…+850 °C, 1,5×100 mm
Artikelnr. 620-0333
Anbieter:   COMARK
Thermometer probes
Artikelnr. 620-0333
Anbieter:  COMARK
Thermometer probes

Mineral insulated semi flexible type K probe, with sub-miniature connector, AK21M, −100…+850 °C, 1,5×100 mm

Preis
310,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Range:
    −100…+850 °C
  • Description:
    Mineral insulated semi flexible type K probe, with sub-miniature connector, AK21M
  • Application:
    General purpose air
  • Ø×L:
    1,5×100 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.