Orders LinkContactUsLinkComponent
Horizontal developing chambers
Horizontal developing chambers
Chamber for 100×100 mm plates
Catalog # 552-0047
Supplier:   CAMAG
Horizontal developing chambers
Catalog # 552-0047
Supplier:  CAMAG
Horizontal developing chambers
Horizontal developing chambers
Chamber for 100×100 mm plates
Price
1 210,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Horizontal developing chambers allows the number of samples to be doubled when compared to a vertical tank. Plates are developed from opposite sides simultaneously towards the middle.
  • Choice of two configurations: sandwich or tank
  • Suitable for all types of solvents
  • Available in two plates sizes
Specifications
  • Description:
    Chamber for 100×100 mm plates