Orders LinkContactUsLinkComponent
Flat bottom chambers
Flat bottom chambers
Chamber for 100×100 mm plates, with stainless steel lid
  552-0009
 :   CAMAG
Flat bottom chambers
  552-0009
 :  CAMAG
Flat bottom chambers
Flat bottom chambers
Chamber for 100×100 mm plates, with stainless steel lid
196,00 €
1 * 1 ST
These flat bottom chamber allow the chromatogram to be developed in a tank atmosphere of partial or complete saturation with solvent vapours. The degree of layer pre-saturation cannot be controlled unless additional accessories are used.
  • Description:
    Chamber for 100×100 mm plates, with stainless steel lid
  • Seq:
    04
Accessories
  •  
    Description
  • 552-0079
    Saturation pads
    1 * 100 ST - 245,00 €