Order Entry
Austria
Orders LinkContactUsLinkComponent
Mesitylene 99%, extra pure

Mesitylene 99%, extra pure

Glass bottle, 250 mL
Catalog # ACRO125582500
Irritant
Environment
Flammable
Health
Mesitylene 99%, extra pure
Catalog # ACRO125582500
CAS Number:  108-67-8
Mesitylene 99%, extra pure
Mesitylene 99%, extra pure

Glass bottle, 250 mL

CAS Number:  108-67-8
Price
56,40 €
Log in for availability
1 * 250 mL
Product Details & Documents
Synonyms:  1,3,5-Trimethylbenzene
  • CAS Number:108-67-8
  • Formula:C₉H₁₂
  • MDL Number:MFCD00008538
  • Molecular Weight:120.19 g/mol
  • Merck Index:13,05934
  • Boiling Pt:163…166 °C (760 mmHg)
  • Melting Pt:–45 °C
  • Density:0.8675 g/cm³
  • Flash Pt:44 °C
  • Storage Temperature:Ambient
  • EINECS:203-604-4
  • UN:2325
  • ADR:3,III

Documents

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4980 to 1.5000 (20°C, 589 nm)
  • GC:
    ≥98.5 %
  • Residue after evaporation:
    ≤0.01 %

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 mL
  • Cat. No.:
    ACRO125582500

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.