2',5'-Dihydroxyacetophenone 97%
Glass bottle for solids, 5 g
Supplier: Thermo Fisher Scientific
2',5'-Dihydroxyacetophenone 97%
Catalog # ACRO114810050
Supplier: Thermo Fisher Scientific
CAS Number: 490-78-8
Jump to:
Product Details & Documents
Synonyms: 1-(2,5-Dihydroxyphenyl)ethan-1-one, 1-(2,5-Dihydroxyphenyl)ethanone, 2,5-Dihydroxyacetophenone
- CAS Number:490-78-8
- Formula:C₈H₈O₃
- MDL Number:MFCD00002343
- Molecular Weight:152.15 g/mol
- Boiling Pt:296 °C (1013 hPa)
- Melting Pt:203…208 °C
- EINECS:207-716-4
Specifications
Specification Test Results
- Appearance:Yellow to yellow-green Crystalline powder or crystals
- Infrared spectrum:Conforms
- Melting point:203 to 208 °C (Dec)
- HPLC:≥96.0 %
- Clarity of solution:passes test
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g



