Orders LinkContactUsLinkComponent
Hydrometers, Brix
Hydrometers, Brix
30 - 60 °Bx, 0,1 °Bx, 300 mm
Catalog # ALLA3910FC030/20QP
Supplier:   Alla France
Hydrometers, Brix
Catalog # ALLA3910FC030/20QP
Supplier:  Alla France
Hydrometers, Brix
Hydrometers, Brix
30 - 60 °Bx, 0,1 °Bx, 300 mm
Price
38,50 €
Log in for availability
1 * 1 ST
Product Details & Documents
These Brix sugar scale hydrometers are specially designed for use in breweries.
Specifications
  • Length:
    300 mm
  • Division:
    0,1 °Bx
  • Range:
    30 - 60 °Bx