Baumé hydrometers
60 - 70, 0,1, 300 mm
ALLA0100FG070/15QP
Baumé hydrometers calibrated at 15 °C.
- Length:300 mm
- Seq:26
- Division:0,1
- Range:60 - 70
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



