Orders LinkContactUsLinkComponent
Spider evaporators | Avantor
Spider evaporators
Spider with evaporating flasks, SJ29/32, glass, 50 ml, For simultaneous distillation from 5×50 ml evaporating flasks with STJ 24/29
Catalog # 531-1319
Supplier:   BUCHI
Spider evaporators
Catalog # 531-1319
Supplier:  BUCHI
Spider evaporators
Spider evaporators
Spider with evaporating flasks, SJ29/32, glass, 50 ml, For simultaneous distillation from 5×50 ml evaporating flasks with STJ 24/29
Price
1 573,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Spider evaporators are ideal when users need to concentrate several samples under the same conditions.
  • Models for distillation with evaporating or cylindrical flasks available
  • Parallel distillation saves time
Specifications
  • Description:
    Spider with evaporating flasks, SJ29/32, glass, 50 ml
  • Type:
    For simultaneous distillation from 5×50 ml evaporating flasks with STJ 24/29
Empfehlungen
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen.