Evaporating flasks
50 ml, 24/29 NS, uncoated
Catalog # BUCH000472
Supplier: BUCHI
Jump to:
Product Details & Documents
Borosilicate glass.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
For Rotavapor® rotary evaporators. Centre neck. Pear-shaped with ground joint socket or EL-flange.
Specifications
- Socket size:24/29 NS
- Type:uncoated
- Capacity:50 ml
- Seq:01