Orders LinkContactUsLinkComponent
Receiving flasks
Receiving flasks
2000 ml, uncoated, KS 35/20
Catalog # 531-1836
Supplier:   BUCHI
Receiving flasks
Catalog # 531-1836
Supplier:  BUCHI
Receiving flasks
Receiving flasks
2000 ml, uncoated, KS 35/20
Price
241,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Especially for BÜCHI rotary evaporators. With spherical joint socket KS 35/20.
Specifications
  • Socket:
    KS 35/20
  • Type:
    uncoated
  • Capacity:
    2000 ml